Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(3-Chloro-5-(trifluoromethoxy)phenyl)propionic acid ≥94%

Supplier: Alfa Aesar
Warning

ALFAH31679.03EA 466 SGD InStock
ALFAH31679.03 ALFAH31679.MD
3-(3-Chloro-5-(trifluoromethoxy)phenyl)propionic acid ≥94%
3-(3-Chloro-5-(trifluoromethoxy)phenyl)propionic acid


Formula: C₁₀H₈ClF₃O₃
MW: 268,62
Storage Temperature: Ambient
MDL Number: MFCD09025398
CAS Number: 916420-74-1
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty