Ethyl (2-fluorophenyl)acetate ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Ethyl 2-fluorophenylacetate
ALFAL19999.14EA
0
SGD
ALFAL19999.14
ALFAL19999.06
Ethyl (2-fluorophenyl)acetate ≥99%
Ethyl (2-fluorophenyl)acetate
Formula:
C₁₀H₁₁FO₂ MW: 182,20 g/mol Boiling Pt: 235 °C Density: 1,101 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04039343 CAS Number: 584-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...