Eppendorf ThermoMixer® F1.5 is customised for 1,5 ml reaction vessels and Eppendorf ThermoMixer® FP is designed for plates. Units feature simple and intuitive operation using predefined temperature keys (37, 42, 56 and 95 °C). Both units are ideal for bacterial growth, enzymatic reactions and lysis reactions at 100 °C. The F1.5 is suitable for denaturation of DNA, RNA and proteins, labelling of nucleic acids and proteins and Proteinase K digestions of cells and tissues. The FP model is suitable for resuspending pellets, mixing protein quantification assays and ELISA assays.
- Precise temperature control from ambient +4 to 100 °C
- Efficient mixing up to 1500 min⁻¹ ( F1.5) or 2000 min⁻¹ (FP) thanks to 2DMix-Control and anti-spill technology
- Digital display enables simple and accurate setting and viewing of parameters
- Prevent condensation on tube lids and walls and improve temperature homogeneity with the new ThermoTop®
Reliable prevention of condensate formation in small vessels can be achieved with the heated lid Eppendorf ThermoTop (optional).
Ordering information: Eppendorf ThermoMixer® F1.5 is supplied with fixed SmartBlock™ for 24×1,5 ml reaction vessels, and Eppendorf ThermoMixer® FP is supplied with fixed SmartBlock™ for microplates and deepwell plates, including lid. Can be used with ThermoTop® cover.