Disposable particle filtering respirators, FFP1/FFP2, 8300 series
Supplier: 3M
MMMA8320EA 52 SGD
MMMA8320 MMMA8310
Disposable particle filtering respirators, FFP1/FFP2, 8300 series
Lightweight respirators with robust outer shell and braided headband offer high durability. Ideal for paint, construction, cement and metal working industries.
Lightweight respirators with robust outer shell and braided headband offer high durability. Ideal for paint, construction, cement and metal working industries.
  • Breathability thanks to 3M™ Advanced Electret filter material
  • Cool Flow™ valve on models “with valve” reduces heat and moisture build-up to offer workers comfortable protection, even in hot and humid conditions (only for 8312 and 8322)
  • Noseclip is quick and easy for wearers to mould around the nose offering greater comfort
  • Colour coded headbands offer easy protection level identification (FFP1 = yellow; FFP2 = blue; FFP3 = red)
  • Flexible, textured edges, super-soft, cushioned inner lining and braided headband for a comfortable and secure fit
  • Breathability thanks to 3M™ Advanced Electret filter material
  • Cool Flow™ valve on models “with valve” reduces heat and moisture build-up to offer workers comfortable protection, even in hot and humid conditions (only for 8312 and 8322)
  • Noseclip is quick and easy for wearers to mould around the nose offering greater comfort
  • Colour coded headbands offer easy protection level identification (FFP1 = yellow; FFP2 = blue; FFP3 = red)
  • Flexible, textured edges, super-soft, cushioned inner lining and braided headband for a comfortable and secure fit
EN 149:2001 + A1:2009 FFP1/FFP2 NR D
Certifications: EN 149:2001 + A1:2009 FFP1/FFP2 NR D
Caution: * One size fits all
* One size fits all