5-Indoleboronic acid 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(1H-indol-5-yl)boronic acid
, 5-Indolylboronic acid
, (1H-Indol-5-yl)boronic acid
ALFAH27008.03EA
199
SGD
ALFAH27008.03
5-Indoleboronic acid 95%
5-Indoleboronic acid
Formula:
C₈H₈BNO₂ MW: 160,97 g/mol Melting Pt: 170…175 °C |
MDL Number:
MFCD01319013 CAS Number: 144104-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...