2-Fluoro-(4-methoxycarbonyl)phenylboronic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Fluoro-4-(methoxycarbonyl)phenylboronic acid
, 2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid
ALFAH52971.03EA
179
SGD
ALFAH52971.03
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid ≥98%
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid
Formula:
C₈H₈BFO₄ MW: 197,96 g/mol Melting Pt: 173…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD08436034 CAS Number: 603122-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...