3-Methoxy-4-(trifluoromethyl)benzamide ≥97%
|
Warning
|
ALFAH33121.06EA
763
SGD
ALFAH33121.06
ALFAH33121.03
3-Methoxy-4-(trifluoromethyl)benzamide ≥97%
3-Methoxy-4-(trifluoromethyl)benzamide
Formula:
C₉H₈F₃NO₂ MW: 219,16 g/mol Melting Pt: 219…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD13185679 CAS Number: 1214328-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...