4-Chloro-3-ethoxy-2-fluorobenzylamine 97%
|
Danger
|
ALFAH33498.03EA
409
SGD
ALFAH33498.03
ALFAH33498.MD
4-Chloro-3-ethoxy-2-fluorobenzylamine 97%
4-Chloro-3-ethoxy-2-fluorobenzylamine
Formula:
C₉H₁₁ClFNO MW: 203,64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19687221 CAS Number: 1323966-28-4 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...