4-Chloro-2,5-xylidine ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Chloro-2,5-dimethylaniline
ALFAH26018.06EA
0
SGD
ALFAH26018.06
4-Chloro-2,5-xylidine ≥98%
4-Chloro-2,5-xylidine
Formula:
C₈H₁₀ClN MW: 155,63 g/mol Melting Pt: 91…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD08445705 CAS Number: 20782-94-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...