2-Methoxy-4-(trifluoromethyl)benzylamine 97%
|
Danger
|
ALFAH32832.03EA
302
SGD
ALFAH32832.03
ALFAH32832.MD
2-Methoxy-4-(trifluoromethyl)benzylamine 97%
2-Methoxy-4-(trifluoromethyl)benzylamine
Formula:
C₉H₁₀F₃NO MW: 205,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06213016 CAS Number: 771581-99-8 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...