3-Methoxy-5-(trifluoromethyl)benzylamine 97%
|
Danger
|
ALFAH32782.03EA
272
SGD
ALFAH32782.03
ALFAH32782.MD
3-Methoxy-5-(trifluoromethyl)benzylamine 97%
3-Methoxy-5-(trifluoromethyl)benzylamine
Formula:
C₉H₁₀F₃NO MW: 205,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09025412 CAS Number: 916420-96-7 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...