4-Methoxy-2-(trifluoromethyl)benzylamine 97%
|
Danger
|
ALFAH32730.06EA
849
SGD
ALFAH32730.06
ALFAH32730.03
4-Methoxy-2-(trifluoromethyl)benzylamine 97%
4-Methoxy-2-(trifluoromethyl)benzylamine
Formula:
C₉H₁₀F₃NO MW: 205,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09025405 CAS Number: 916420-84-3 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...