2-Biphenylboronic acid 95%
Supplier: Thermo Scientific
|
Warning
|
ALFAL17547.03EA
107
SGD
ALFAL17547.03
2-Biphenylboronic acid 95%
2-Biphenylboronic acid
Formula:
C₁₂H₁₁BO₂ MW: 198,03 g/mol Melting Pt: 173…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00136929 CAS Number: 4688-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...