Silicon nitride ≥99.3% (metals basis) ∼90% β-phase

Synonyms: Silicon(IV) nitride

ALFA40379.09EA 0 SGD
ALFA40379.09 ALFA40379.18 ALFA40379.30
Silicon nitride ≥99.3% (metals basis) ∼90% β-phase
Silicon nitride
Formula: N₄Si₃
MW: 140,28 g/mol
Melting Pt: 1900 °C (subl.)
Density: 3,2
Storage Temperature: Ambient
MDL Number: MFCD00011230
CAS Number: 12033-89-5
EINECS: 234-796-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR