3-Isopropoxyphenylboronic acid ≥97%

Warning

Synonyms: 3-Isopropoxybenzeneboronic acid

ALFAH52981.14EA 692 SGD
ALFAH52981.14
3-Isopropoxyphenylboronic acid ≥97%
3-Isopropoxyphenylboronic acid
Formula: C₉H₁₃BO₃
MW: 180,01 g/mol
Melting Pt: 76…81 °C
Storage Temperature: Ambient
MDL Number: MFCD03701512
CAS Number: 216485-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR