5-Fluoro-2-(trifluoromethoxy)benzyl bromide ≥97%
|
Danger
|
ALFAH32201.MDEA
99.5
SGD
ALFAH32201.MD
ALFAH32201.03
5-Fluoro-2-(trifluoromethoxy)benzyl bromide ≥97%
5-Fluoro-2-(trifluoromethoxy)benzyl bromide
Formula:
C₈H₅BrF₄O MW: 273,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11519337 CAS Number: 1092460-88-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...