2-Ethoxy-3,5-difluorobenzylamine 97%, JRD
|
Danger
|
ALFAH32972.MDEA
104
SGD
ALFAH32972.MD
ALFAH32972.03
2-Ethoxy-3,5-difluorobenzylamine 97%, JRD
2-Ethoxy-3,5-difluorobenzylamine
Formula:
C₉H₁₁F₂NO MW: 187,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09258721 CAS Number: 1017779-86-0 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...