3,5-Difluoro-4-methoxybenzylamine 97%
Supplier: Thermo Scientific
|
Danger
|
ALFAH26693.MDEA
0
SGD
ALFAH26693.MD
3,5-Difluoro-4-methoxybenzylamine 97%
3,5-Difluoro-4-methoxybenzylamine
Formula:
C₈H₉F₂NO MW: 173,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115915 CAS Number: 105969-16-2 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...