3',5'-Difluoro-2'-hydroxyacetophenone ≥97%
|
Warning
|
Synonyms:
1-(3,5-Difluoro-2-hydroxyphenyl)ethanone
ALFAH32624.09EA
441
SGD
ALFAH32624.09
ALFAH32624.03
3',5'-Difluoro-2'-hydroxyacetophenone ≥97%
3',5'-Difluoro-2'-hydroxyacetophenone
Formula:
C₈H₆F₂O₂ MW: 172,13 g/mol Melting Pt: 105…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042476 CAS Number: 140675-42-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...