4'-Fluoro-3'-hydroxyacetophenone ≥98%
Supplier: Thermo Scientific
|
Warning
|
ALFAH32965.03EA
0
SGD
ALFAH32965.03
4'-Fluoro-3'-hydroxyacetophenone ≥98%
4'-Fluoro-3'-hydroxyacetophenone
Formula:
C₈H₇FO₂ MW: 154,14 g/mol Melting Pt: 119…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD12545938 CAS Number: 949159-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...