5-Chloro-2-(trifluoromethyl)benzylamine 97%, JRD
|
Danger
|
ALFAH32593.06EA
660
SGD
ALFAH32593.06
ALFAH32593.03
5-Chloro-2-(trifluoromethyl)benzylamine 97%, JRD
5-Chloro-2-(trifluoromethyl)benzylamine
Formula:
C₈H₇ClF₃N MW: 209,60 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09832359 CAS Number: 942319-05-3 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...