2'-Methoxy-5'-(trifluoromethyl)acetophenone ≥97%
|
Warning
|
Synonyms:
1-(2-Methoxy-5-trifluoromethylphenyl)ethanone
ALFAH32652.03EA
179
SGD
ALFAH32652.03
ALFAH32652.06
2'-Methoxy-5'-(trifluoromethyl)acetophenone ≥97%
2'-Methoxy-5'-(trifluoromethyl)acetophenone
Formula:
C₁₀H₉F₃O₂ MW: 218,17 g/mol Melting Pt: 45…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD09832300 CAS Number: 503464-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...