2'-Methyl-5'-(trifluoromethoxy)acetophenone ≥97%
|
Warning
|
ALFAH33470.03EA
204
SGD
ALFAH33470.03
ALFAH33470.MD
2'-Methyl-5'-(trifluoromethoxy)acetophenone ≥97%
2'-Methyl-5'-(trifluoromethoxy)acetophenone
Formula:
C₁₀H₉F₃O₂ MW: 218,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22201058 CAS Number: 1373920-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...