4-Isopropoxy-3-methylphenylboronic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Isopropoxy-3-methylbenzeneboronic acid
ALFAH52862.03EA
189
SGD
ALFAH52862.03
4-Isopropoxy-3-methylphenylboronic acid ≥97%
4-Isopropoxy-3-methylphenylboronic acid
Formula:
C₁₀H₁₅BO₃ MW: 194,04 g/mol Melting Pt: 128…132 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659871 CAS Number: 850568-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...