Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(-)-Menthyl chloroformate ≥90%

Supplier: Alfa Aesar
Danger



Formula: C₁₁H₁₉ClO₂
MW: 218,73
Boiling Pt: 108-109 °C/11mm
Density: 1,025
Flash Pt: 70 °C (158 °F)
MDL Number: MFCD00009694
CAS Number: 14602-86-9
UN: 3277
ADR: 6.1,II

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 25 g ALFAL11739.14
Supplier Article Number:: L11739.14
Glass bottle for solids 5 g ALFAL11739.06
Supplier Article Number:: L11739.06
Expand All / Collapse All

SPECIFICATIONS

Formula weight 218.73
CAS No. 14602-86-9
Purity 95%
Refractive index 1.4580
Storage temperature 2°C to 8°C
Flash point 70°(158°F)
Boiling point 108-109°/11mm
Molecular formula C11H19ClO2
MDL No. MFCD00009694
UN No. 3277
Optical rotation -82° (c=1 in chloroform)
Beilstein reference 2414686
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping