Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-glycine (Boc-Gly-OH)

Supplier: Panreac Química SLU
Synonyms: Boc-Gly-OH



Formula: C₇H₁₃NO₄
MW: 175,2 g/mol
MDL Number: MFCD00002690
CAS Number: 4530-20-5
EINECS: 224-864-5


 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g PANR15A483.1604
Supplier Article Number:: 15A483.1604
Glass bottle for solids 10 g PANR15A483.1605
Supplier Article Number:: 15A483.1605
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty