Diosmin
Supplier: EDQM
Synonyms:
5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one
EDQMY0000095EA
441
SGD
EDQMY0000095
Diosmin
Diosmin
Formula:
C₂₈H₃₂O₁₅ MW: 608,5 g/mol |
MDL Number:
MFCD00009772 CAS Number: 520-27-4 EINECS: 208-289-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...