N(α)-Cbz-D-histidine ≥98%
Supplier: Thermo Scientific
Synonyms:
N(α)-Benzyloxycarbonyl-D-histidine
, Z-D-His-OH
ALFAL09005.03EA
0
SGD
ALFAL09005.03
N(α)-Cbz-D-histidine ≥98%
N(α)-Cbz-D-histidine
Formula:
C₁₄H₁₅N₃O₄ MW: 289,3 g/mol Melting Pt: 161…163 °C |
MDL Number:
MFCD00067702 CAS Number: 67424-93-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...