10,12-Tricosadiynoic acid 96%
Supplier: Thermo Scientific
Synonyms:
10,12-Tricosadiynoicacid
ALFAL10231.06EA
464
SGD
ALFAL10231.06
ALFAL10231.03
10,12-Tricosadiynoic acid 96%
10,12-Tricosadiynoic acid
Formula:
C₂₃H₃₈O₂ MW: 346,56 g/mol Melting Pt: 55…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041683 CAS Number: 66990-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...