(2-(Dimethylamino)ethyl)triphenylphosphonium bromide 98%
Supplier: Thermo Scientific
|
Warning
|
ALFAL00917.18EA
0
SGD
ALFAL00917.18
ALFAL00917.09
(2-(Dimethylamino)ethyl)triphenylphosphonium bromide 98%
(2-(Dimethylamino)ethyl)triphenylphosphonium bromide
Formula:
C₂₂H₂₅BrNP MW: 414,33 g/mol Melting Pt: ∼ 200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011794 CAS Number: 21331-80-6 EINECS: 244-335-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...