Tris[N,N-bis(trimethylsilyl)amide]yttrium(III) ≥98%
|
Danger
|
ampouled under argon
ALFA39578.03EA
277
SGD
ALFA39578.03
ALFA39578.06
Tris[N,N-bis(trimethylsilyl)amide]yttrium(III) ≥98%
Tris[N,N-bis(trimethylsilyl)amide]yttrium (III)
Formula:
C₁₈H₅₄N₃Si₆Y MW: 570,06 g/mol Boiling Pt: 105 °C (10^-4 mmHg) (subl.) Melting Pt: 180…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD00210649 CAS Number: 41836-28-6 UN: 3396 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...