4-Boronobenzenesulphonamide ≥97%

Warning

Synonyms: 4-Sulphamoylbenzeneboronic acid(4-Aminosulphonylphenyl)boronic acid

ALFAH52928.MDEA 65.2 SGD
ALFAH52928.MD ALFAH52928.03
4-Boronobenzenesulphonamide ≥97%
4-Boronobenzenesulphonamide
Formula: C₆H₈BNO₄S
MW: 201,00 g/mol
Melting Pt: 250…256 °C
Storage Temperature: Ambient
MDL Number: MFCD06659839
CAS Number: 613660-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR