4-Chloro-2-formylphenylboronic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Chloro-2-formylbenzeneboronic acid
ALFAH52549.MDEA
124
SGD
ALFAH52549.MD
4-Chloro-2-formylphenylboronic acid ≥98%
4-Chloro-2-formylphenylboronic acid
Formula:
C₇H₆BClO₃ MW: 184,39 g/mol Melting Pt: 132…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD08689492 CAS Number: 913835-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...