3'-Fluoro-5'-(pentafluorothio)acetophenone ≥97%
|
Warning
|
ALFAH33302.03EA
424
SGD
ALFAH33302.03
ALFAH33302.MD
3'-Fluoro-5'-(pentafluorothio)acetophenone ≥97%
3'-Fluoro-5'-(pentafluorothio)acetophenone
Formula:
C₈H₆F₆OS MW: 264,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16652464 CAS Number: 1240257-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...