(1S,2S)-2-Methoxycyclohexanol 97%, ChiPros®
|
Warning
|
Synonyms:
(1S,2S)-(+)-2-Methoxycyclohexanol
ALFAL19065.14EA
827
SGD
ALFAL19065.14
ALFAL19065.03
ALFAL19065.06
(1S,2S)-2-Methoxycyclohexanol 97%, ChiPros®
(1S,2S)-2-Methoxycyclohexanol
Formula:
C₇H₁₄O₂ MW: 130,19 g/mol Boiling Pt: 195 °C Density: 1,015 Storage Temperature: Ambient |
MDL Number:
MFCD00211253 CAS Number: 134108-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...