2,2,2-Trifluoroethyl methyl ether ≥99%
Supplier: Thermo Scientific
|
Danger
|
ALFAL17887.06EA
117
SGD
ALFAL17887.06
ALFAL17887.14
2,2,2-Trifluoroethyl methyl ether ≥99%
2,2,2-Trifluoroethyl methyl ether
Formula:
C₃H₅F₃O MW: 114,07 g/mol Boiling Pt: 30 °C Density: 1,156 Flash Pt: –8 °C (–18 °F) |
MDL Number:
MFCD00236114 CAS Number: 460-43-5 UN: 1993 ADR: 3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...