3-Ethoxy-2,6-difluorophenol 97%

Danger

ALFAH31776.MDEA 48.3 SGD
ALFAH31776.MD ALFAH31776.03
3-Ethoxy-2,6-difluorophenol 97%
3-Ethoxy-2,6-difluorophenol
Formula: C₈H₈F₂O₂
MW: 174,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519373
CAS Number: 1092461-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR