3-Methylhippuric acid ≥97%
Supplier: Thermo Scientific
Synonyms:
2-(3-Methylbenzamido)acetic acid
ALFAH53458.03EA
71.8
SGD
ALFAH53458.03
3-Methylhippuric acid ≥97%
3-Methylhippuric acid
Formula:
C₁₀H₁₁NO₃ MW: 193,20 g/mol Melting Pt: 138…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044399 CAS Number: 27115-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...