Vincristine sulphate

Supplier: EDQM

EDQMV0405000EA 441 SGD
EDQMV0405000
Vincristine sulphate
Vincristine sulphate
Formula: C₄₆H₅₈N₄O₁₄S
MW: 923,04 g/mol
MDL Number: MFCD00084729
CAS Number: 2068-78-2
EINECS: 218-190-0
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR