4-Chloro-2-ethoxyphenylboronic acid ≥98%
|
Warning
|
Synonyms:
4-Chloro-2-ethoxybenzeneboronic acid
ALFAH52861.03EA
140
SGD
ALFAH52861.03
4-Chloro-2-ethoxyphenylboronic acid ≥98%
4-Chloro-2-ethoxyphenylboronic acid
Formula:
C₈H₁₀BClO₃ MW: 200,43 g/mol Melting Pt: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094940 CAS Number: 850568-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...