Rubidium standard solution, 10000 mg/L Rb in dil. nitric acid (from Rb₂CO₃) standard for ICP
Supplier: PanReac AppliChem
PANR775993.1208EA 725 SGD
PANR775993.1208
Rubidium standard solution, 10000 mg/L Rb in dil. nitric acid (from Rb₂CO₃) standard for ICP
Standards Cationic Standards Rubidium Standards
Certifications: Traceable to NIST.