Hydrocortisone-21-acetate

Supplier: EDQM

Synonyms: Hydrocortisone acetate

EDQMY0001482EA 419 SGD
EDQMY0001482
Hydrocortisone-21-acetate
Hydrocortisone 21-acetate
Formula: C₂₃H₃₂O₆
MW: 404,51 g/mol
MDL Number: MFCD00037714
CAS Number: 50-03-3
EINECS: 200-004-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR