2,6-Dimethoxy-4-methylbenzaldehyde ≥97%

Supplier: Thermo Scientific
Warning

ALFAH26982.09EA 567 SGD
ALFAH26982.09 ALFAH26982.03
2,6-Dimethoxy-4-methylbenzaldehyde ≥97%
2,6-Dimethoxy-4-methylbenzaldehyde
Formula: C₁₀H₁₂O₃
MW: 180,20 g/mol
Melting Pt: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD01321089
CAS Number: 6937-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR