Flow cup viscosimeters "Ford"
Supplier: Fungilab
FUNGFCA003EA 811 SGD
FUNGFCA003 FUNGFCA002 FUNGFCA004
Flow cup viscosimeters "Ford"
Viscometers Flow Cup Viscometers
Anodised aluminium cups with fixed stainless steel nozzles.
  • Rapid measurement and comparison of viscosities in the paint, ink and varnishing industries
  • Calibration certificates at specified temperatures included with the cups
  • Equations for relating flow times to viscosity
Designed for the measurement of kinematic viscosity by comparison of the break-thread flow times of similar liquids.
Certifications: ASTM D1200
Ordering information: Please order the stand and calibration fluids separately.