4-(1-Naphthyl)phenylboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-(1-Naphthyl)benzeneboronic acid

ALFAH52710.03EA 0 SGD
ALFAH52710.03 ALFAH52710.MD
4-(1-Naphthyl)phenylboronic acid ≥97%
4-(1-Naphthyl)phenylboronic acid
Formula: C₁₆H₁₃BO₂
MW: 248,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669639
CAS Number: 870774-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR