6-Hydroxy-2-naphthaleneboronic acid ≥96%
|
Warning
|
Synonyms:
(6-Hydroxynaphthalen-2-yl)boronic acid
ALFAH53074.03EA
335
SGD
ALFAH53074.03
6-Hydroxy-2-naphthaleneboronic acid ≥96%
6-Hydroxy-2-naphthaleneboronic acid
Formula:
C₁₀H₉BO₃ MW: 187,99 g/mol Melting Pt: 218…224 °C Storage Temperature: Ambient |
MDL Number:
MFCD00092986 CAS Number: 173194-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...