(4-Methylphenyl)methanesulphonyl chloride ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: p-Tolylmethanesulphonyl chloride4-Methylbenzylsulphonyl chlorideα-p-Xylenesulphonyl chloride

ALFAH26038.03EA 0 SGD
ALFAH26038.03
(4-Methylphenyl)methanesulphonyl chloride ≥97%
(4-Methylphenyl)methanesulphonyl chloride
Formula: C₈H₉ClO₂S
MW: 204,67 g/mol
Melting Pt: 75…81 °C
Storage Temperature: Ambient
MDL Number: MFCD01631931
CAS Number: 51419-59-1
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR