5-Chloro-2-(trifluoromethoxy)benzyl alcohol ≥97%
|
Warning
|
ALFAH32806.03EA
272
SGD
ALFAH32806.03
ALFAH32806.MD
5-Chloro-2-(trifluoromethoxy)benzyl alcohol ≥97%
5-Chloro-2-(trifluoromethoxy)benzyl alcohol
Formula:
C₈H₆ClF₃O₂ MW: 226,58 g/mol Melting Pt: 52…54 °C Storage Temperature: Ambient |
MDL Number:
MFCD11519357 CAS Number: 874821-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...