4-Methoxy-3-(trifluoromethoxy)benzyl alcohol ≥97%

Warning

ALFAH33588.03EA 218 SGD
ALFAH33588.03 ALFAH33588.MD
4-Methoxy-3-(trifluoromethoxy)benzyl alcohol ≥97%
4-Methoxy-3-(trifluoromethoxy)benzyl alcohol
Formula: C₉H₉F₃O₃
MW: 221,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18398472
CAS Number: 853771-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR