3-Methoxy-4-(trifluoromethyl)benzyl alcohol ≥97%
|
Warning
|
ALFAH33093.06EA
624
SGD
ALFAH33093.06
ALFAH33093.03
3-Methoxy-4-(trifluoromethyl)benzyl alcohol ≥97%
3-Methoxy-4-(trifluoromethyl)benzyl alcohol
Formula:
C₉H₉F₃O₂ MW: 206,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18398590 CAS Number: 276861-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...